Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGACT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075571.1 | NP_001069039.1 | 2 - 3 | 146 | 72 | |
GenBank mRNA | BC118322.1 | - | 2 - 3 | 146 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |