Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075580.2 | NP_001069048.1 | 1 - 2 | 63 | 107 | |
GenBank mRNA | BC120123.1 | - | 1 - 2 | 64 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |