Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075626.1 | NP_001069094.1 | 4 - 5 | 620 | 74 | |
GenBank mRNA | BC120092.1 | - | 4 - 5 | 620 | 74 | |
BT029881.1 | - | 4 - 5 | 631 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |