Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075638.2 | NP_001069106.1 | 4 - 5 | 308 | 104 | |
GenBank mRNA | BC118351.1 | - | 4 - 5 | 309 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |