Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075769.1 | NP_001069237.1 | 10 - 11 | 1303 | 152 | |
GenBank mRNA | BC123609.1 | - | 10 - 11 | 1303 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |