Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075815.1 | NP_001069283.1 | 7 - 8 | 1273 | 74 | |
GenBank mRNA | BC119954.1 | - | 7 - 8 | 1273 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |