Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075917.1 | NP_001069385.1 | 4 - 5 | 576 | 83 | |
GenBank mRNA | BC120349.1 | - | 4 - 5 | 576 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |