Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075917.1 | NP_001069385.1 | 5 - 6 | 732 | 65 | |
GenBank mRNA | BC120349.1 | - | 5 - 6 | 732 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |