Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075940.2 | NP_001069408.1 | 3 - 4 | 844 | 60 | |
GenBank mRNA | BC120182.1 | - | 3 - 4 | 659 | 60 | |
BT025409.1 | - | 3 - 4 | 609 | 60 | ||
BT025454.1 | - | 3 - 4 | 636 | 60 | ||
BT026254.1 | - | 3 - 4 | 650 | 60 | ||
BT026255.1 | - | 3 - 4 | 844 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |