Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC120308.1 | - | 3 - 4 | 589 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |