Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075988.2 | NP_001069456.1 | 3 - 4 | 555 | 109 | |
GenBank mRNA | BC120264.1 | - | 3 - 4 | 556 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |