Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075990.1 | NP_001069458.1 | 11 - 12 | 1112 | 103 | |
GenBank mRNA | BC118307.1 | - | 11 - 12 | 1112 | 103 | |
BT025484.1 | - | 11 - 12 | 1095 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |