Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076007.2 | NP_001069475.1 | 2 - 3 | 187 | 59 | |
GenBank mRNA | BC120280.1 | - | 2 - 3 | 188 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |