Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076009.1 | NP_001069477.1 | 4 - 5 | 1100 | 107 | |
GenBank mRNA | BC120040.1 | - | 4 - 5 | 1100 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |