Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076097.1 | NP_001069565.1 | 1 - 2 | 119 | 116 | |
GenBank mRNA | BC109942.1 | - | 1 - 2 | 119 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |