Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMKLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076113.2 | NP_001069581.1 | 1 - 2 | 166 | 96 | |
GenBank mRNA | BC120203.1 | - | 1 - 2 | 167 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |