Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMKLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076113.2 | NP_001069581.1 | 3 - 4 | 628 | 85 | |
GenBank mRNA | BC120203.1 | - | 3 - 4 | 629 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |