Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076194.1 | NP_001069662.1 | 2 - 3 | 416 | 98 | |
GenBank mRNA | BT026338.1 | - | 2 - 3 | 416 | 98 | |
KU253656.1 | - | 2 - 3 | 294 | 98 | ||
KU253657.1 | - | 2 - 3 | 397 | 98 | ||
KU253658.1 | - | 2 - 3 | 397 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |