Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076212.2 | NP_001069680.1 | 2 - 3 | 821 | 102 | |
GenBank mRNA | BC120277.1 | - | 2 - 3 | 822 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |