Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076212.2 | NP_001069680.1 | 3 - 4 | 948 | 64 | |
GenBank mRNA | BC120277.1 | - | 3 - 4 | 949 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |