Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076817.1 | NP_001070285.1 | 10 - 11 | 1421 | 99 | |
GenBank mRNA | BC123865.1 | - | 10 - 11 | 1421 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |