Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076817.1 | NP_001070285.1 | 11 - 12 | 1641 | 124 | |
GenBank mRNA | BC123865.1 | - | 11 - 12 | 1641 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |