Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076831.1 | NP_001070299.1 | 2 - 3 | 363 | 79 | |
GenBank mRNA | BC123469.1 | - | 2 - 3 | 363 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |