Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077946.1 | NP_001071414.2 | 6 - 7 | 881 | 66 | |
GenBank mRNA | BC122770.1 | - | 6 - 7 | 881 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |