Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077988.2 | NP_001071456.1 | 1 - 2 | 234 | 92 | |
GenBank mRNA | BC123576.1 | - | 1 - 2 | 235 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |