Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078014.1 | NP_001071482.1 | 2 - 3 | 718 | 81 | |
GenBank mRNA | BC119905.1 | - | 2 - 3 | 718 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |