Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079627.2 | NP_001073095.1 | 4 - 5 | 338 | 80 | |
GenBank mRNA | BC120168.1 | - | 4 - 5 | 340 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |