Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM210A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080259.1 | NP_001073728.1 | 1 - 2 | 95 | 129 | |
GenBank mRNA | BC122695.1 | - | 1 - 2 | 95 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |