Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080338.1 | NP_001073807.1 | 2 - 3 | 355 | 151 | |
GenBank mRNA | BC122676.1 | - | 2 - 3 | 355 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |