Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080340.2 | NP_001073809.1 | 5 - 6 | 928 | 91 | |
GenBank mRNA | BC120149.1 | - | 5 - 6 | 929 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |