Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080902.2 | NP_001074371.2 | 1 - 2 | 134 | 151 | |
GenBank mRNA | BC148916.1 | - | 1 - 2 | 132 | 151 | |
BT026154.1 | - | 1 - 2 | 129 | 151 | ||
EH146650.1 | - | 1 - 2 | 135 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |