Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173893.3 | NP_776318.1 | 1 - 2 | 106 | 76 | |
GenBank mRNA | BC118352.1 | - | 1 - 2 | 106 | 76 |
Gene Symbol: LOC783680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB098890.1 | - | 1 - 2 | 89 | 76 | |
AY911322.1 | - | 1 - 2 | 98 | 76 | ||
X69084.1 | - | 79 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Endogenous Controls Bos taurus Inventoried Probe spans exons |