Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003903.3 | NP_001003903.1 | 2 - 3 | 340 | 112 | |
GenBank mRNA | AY682746.1 | - | 2 - 3 | 281 | 112 | |
BC133442.1 | - | 2 - 3 | 342 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |