Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014845.3 | NP_001014845.1 | 3 - 4 | 576 | 74 | |
GenBank mRNA | BC133432.1 | - | 3 - 4 | 577 | 74 | |
EU935419.1 | - | 3 - 4 | 371 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |