Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024524.3 | NP_001019695.2 | 7 - 8 | 1041 | 112 | |
GenBank mRNA | BC126551.1 | - | 7 - 8 | 1042 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |