Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040475.2 | NP_001035565.2 | 1 - 2 | 69 | 55 | |
GenBank mRNA | BC109815.1 | - | 1 - 2 | 69 | 55 | |
DT851934.1 | - | 1 - 2 | 69 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |