Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040549.2 | NP_001035639.1 | 2 - 3 | 375 | 102 | |
GenBank mRNA | BC123529.1 | - | 2 - 3 | 375 | 102 | |
BT021049.1 | - | 2 - 3 | 376 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |