Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076834.1 | NP_001070302.1 | 1 - 2 | 132 | 130 | |
GenBank mRNA | BC123820.1 | - | 1 - 2 | 168 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |