Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077486.2 | NP_001070954.1 | 2 - 3 | 876 | 72 | |
GenBank mRNA | AB238936.1 | - | 1 - 2 | 559 | 72 | |
AF245487.1 | - | 1 - 2 | 409 | 72 | ||
AF245488.1 | - | 1 - 2 | 409 | 72 | ||
AF245489.1 | - | 1 - 2 | 409 | 72 | ||
BC133281.1 | - | 2 - 3 | 876 | 72 | ||
BC147863.1 | - | 2 - 3 | 1011 | 72 | ||
HM630302.1 | - | 1 - 2 | 576 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |