Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077856.1 | NP_001071324.1 | 2 - 3 | 216 | 75 | |
GenBank mRNA | BC126760.1 | - | 2 - 3 | 216 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |