Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077858.2 | NP_001071326.1 | 3 - 4 | 446 | 83 | |
GenBank mRNA | BC126649.1 | - | 3 - 4 | 448 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |