Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077858.2 | NP_001071326.1 | 9 - 10 | 1121 | 76 | |
GenBank mRNA | BC126649.1 | - | 9 - 10 | 1123 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |