Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077924.2 | NP_001071392.1 | 5 - 6 | 615 | 74 | |
GenBank mRNA | BC126573.1 | - | 5 - 6 | 616 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |