Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077965.2 | NP_001071433.1 | 3 - 4 | 393 | 66 | |
GenBank mRNA | BC126700.1 | - | 3 - 4 | 395 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |