Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077998.2 | NP_001071466.1 | 7 - 8 | 1088 | 87 | |
GenBank mRNA | BC126545.1 | - | 7 - 8 | 1089 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |