Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079580.2 | NP_001073048.1 | 4 - 5 | 705 | 87 | |
GenBank mRNA | BC122771.1 | - | 4 - 5 | 706 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |