Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079646.1 | NP_001073114.1 | 3 - 4 | 128 | 69 | |
GenBank mRNA | BC126739.1 | - | 3 - 4 | 128 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |