Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080216.1 | NP_001073685.1 | 6 - 7 | 997 | 90 | |
GenBank mRNA | BT029866.1 | - | 6 - 7 | 997 | 90 | |
D16639.1 | - | 8 - 9 | 897 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |