Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080223.1 | NP_001073692.1 | 3 - 4 | 430 | 62 | |
GenBank mRNA | BC126741.1 | - | 3 - 4 | 430 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |