Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080238.1 | NP_001073707.1 | 1 - 2 | 726 | 131 | |
GenBank mRNA | BC146065.1 | - | 4 - 5 | 873 | 131 | |
BT029783.1 | - | 1 - 2 | 726 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |